accessibility menu, dialog, popup

UserName

CAS RN 16289-95-5

IUPAC Name: 2,2'-dihydroxy-1H,1'H,2H,2'H,3H,3'H-[2,2'-biindene]-1,1',3,3'-tetrone dihydrate
Synonyms: hydrindantin dihydrate
Molecular Weight (g/mol): 358.3
Molecular Formula: C18H14O8
InChi Key: QIAIXVRAKYAOGJ-UHFFFAOYSA-N
SMILES: O.O.OC1(C(=O)C2=CC=CC=C2C1=O)C1(O)C(=O)C2=CC=CC=C2C1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results