CAS RN 162157-03-1
IUPAC Name:
λ²-iron(2+) 2-(cyclopenta-2,4-dien-1-ylidene)-4-(propan-2-yl)-1,3-oxazolidin-3-ide cyclopenta-2,4-dien-1-ide
Synonyms:
λ²-iron(2+) 2-(cyclopenta-2,4-dien-1-ylidene)-4-isopropyl-1,3-oxazolidin-3-ide cyclopentadienide
Molecular Weight (g/mol):
297.18
Molecular Formula:
C16H19FeNO
InChi Key:
MOTUFOCNRSDOMK-UHFFFAOYNA-N
SMILES:
[Fe++].[CH-]1C=CC=C1.CC(C)C1COC([N-]1)=C1C=CC=C1
1
–
1
of
1
results
1
(S)-(4-Isopropyloxazolin-2-yl)ferrocene 98.0+%, TCI America™
CAS: 162157-03-1 Molecular Formula: C16H19FeNO Molecular Weight (g/mol): 297.179 InChI Key: MOTUFOCNRSDOMK-YQFADDPSSA-N PubChem CID: 70700147 IUPAC Name: cyclopenta-1,3-diene;(4S)-2-cyclopenta-2,4-dien-1-ylidene-4-propan-2-yl-1,3-oxazolidin-3-ide;iron(2+) SMILES: CC(C)C1COC(=C2C=CC=C2)[N-]1.[CH-]1C=CC=C1.[Fe+2]