missing translation for 'onlineSavingsMsg'
Learn More

(S)-(4-Isopropyloxazolin-2-yl)ferrocene 98.0+%, TCI America™

Catalog No. I07491G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I07491G Supplier TCI America Supplier No. I07491G

Chemical Identifiers

CAS 162157-03-1
Molecular Formula C16H19FeNO
Molecular Weight (g/mol) 297.179
InChI Key MOTUFOCNRSDOMK-YQFADDPSSA-N
PubChem CID 70700147
IUPAC Name cyclopenta-1,3-diene;(4S)-2-cyclopenta-2,4-dien-1-ylidene-4-propan-2-yl-1,3-oxazolidin-3-ide;iron(2+)
SMILES CC(C)C1COC(=C2C=CC=C2)[N-]1.[CH-]1C=CC=C1.[Fe+2]

Specifications

Quantity 1 g
Formula Weight 297.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(4-Isopropyloxazolin-2-yl)ferrocene