accessibility menu, dialog, popup

UserName

NOHOOHOBr

CAS RN 162102-81-0

IUPAC Name: 4-bromopyridine-2,6-dicarboxylic acid
Synonyms: 4-bromopyridine-2,6-dicarboxylic acid
Molecular Weight (g/mol): 246.02
Molecular Formula: C7H4BrNO4
InChi Key: JVCWTTKJXQPITQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Br)=CC(=N1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results