missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,6-pyridinedicarboxylic Acid Monohydrate 98.0+%, TCI America™

Catalog No. B4970200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4970200MG 200 mg
B49701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4970200MG Supplier TCI America Supplier No. B4970200MG
Only null left

Chemical Identifiers

CAS 162102-81-0
Molecular Formula C7H4BrNO4
Molecular Weight (g/mol) 246.016
MDL Number MFCD09033573
InChI Key JVCWTTKJXQPITQ-UHFFFAOYSA-N
PubChem CID 15832860
IUPAC Name 4-bromopyridine-2,6-dicarboxylic acid
SMILES C1=C(C=C(N=C1C(=O)O)C(=O)O)Br

Specifications

Melting Point 206°C
Color White
Quantity 200 mg
Formula Weight 246.02
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2,6-pyridinedicarboxylic Acid Monohydrate