CAS RN 1620086-77-2
CAS RN 1620086-77-2
IUPAC Name:
6-chloro-N,N-bis(propan-2-yl)-2,4-dihydro-1,3,2-benzodioxaphosphinin-2-amine
Synonyms:
6-chloro-N,N-diisopropyl-4H-1,3,2-benzodioxaphosphinin-2-amine
Molecular Weight (g/mol):
287.72
Molecular Formula:
C13H19ClNO2P
InChi Key:
ZJPWJYRCWIYHMH-UHFFFAOYNA-N
SMILES:
CC(C)N(C(C)C)P1OCC2=C(O1)C=CC(Cl)=C2
1
–
1
of
1
results
1
5-Chlorosaligenyl-N,N-diisopropylphosphoramidite 95.0+%, TCI America™
CAS: 1620086-77-2 Molecular Formula: C13H19ClNO2P Molecular Weight (g/mol): 287.724 InChI Key: ZJPWJYRCWIYHMH-UHFFFAOYSA-N Synonym: 6-Chloro-N,N-diisopropyl-4H-benzo[d][1,3,2]dioxaphosphinin-2-amine PubChem CID: 90306628 IUPAC Name: 6-chloro-N,N-di(propan-2-yl)-4H-1,3,2-benzodioxaphosphinin-2-amine SMILES: CC(C)N(C(C)C)P1OCC2=C(O1)C=CC(=C2)Cl