missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chlorosaligenyl-N,N-diisopropylphosphoramidite 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 45°C |
| Color | White |
| Quantity | 200 mg |
| Formula Weight | 287.72 |
| Percent Purity | ≥95.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 5-Chlorosaligenyl-N,N-diisopropylphosphoramidite |
Safety and Handling
| TSCA | No |