accessibility menu, dialog, popup

UserName

CAS RN 161948-70-5

IUPAC Name: 2-({1-[(tert-butoxy)carbonyl]piperidin-4-yl}oxy)acetate
Synonyms: 2-{[1-(tert-butoxycarbonyl)piperidin-4-yl]oxy}acetate
Molecular Weight (g/mol): 258.3
Molecular Formula: C12H20NO5
InChi Key: YHUAHIMRWSVXCN-UHFFFAOYSA-M
SMILES: CC(C)(C)OC(=O)N1CCC(CC1)OCC([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results