accessibility menu, dialog, popup

UserName

CAS RN 16156-59-5

IUPAC Name: phenyl methanesulfonate
Synonyms: phenylmethanesulfonate
Molecular Weight (g/mol): 172.2
Molecular Formula: C7H8O3S
InChi Key: WXVUCMFEGJUVTN-UHFFFAOYSA-N
SMILES: CS(=O)(=O)OC1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)