missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Methanesulfonate 98.0+%, TCI America™

Catalog No. P18525G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P18525G Supplier TCI America Supplier No. P18525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16156-59-5
Molecular Formula C7H8O3S
Molecular Weight (g/mol) 172.20
MDL Number MFCD00095143
InChI Key WXVUCMFEGJUVTN-UHFFFAOYSA-N
Synonym phenyl methylsulfonate, phenyl methyl sulfonate, acmc-1bs7l, phenylmethanesulfonate 4, phenyl methanesulfonate, methanesulfonic acid,phenyl ester, methanesulfonic acid phenyl ester, methanesulfonic acid, phenyl ester
PubChem CID 316170
IUPAC Name phenyl methanesulfonate
SMILES CS(=O)(=O)OC1=CC=CC=C1

Specifications

Melting Point 61°C
Color White-Yellow
Boiling Point 279°C
Quantity 5 g
Formula Weight 172.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Phenyl Methanesulfonate