accessibility menu, dialog, popup

UserName

ClNHBrNCl

CAS RN 16076-27-0

IUPAC Name: 2-bromo-4,5-dichloro-1H-imidazole
Synonyms: 2-bromo-4,5-dichloro-1H-imidazole
Molecular Weight (g/mol): 215.86
Molecular Formula: C3HBrCl2N2
InChi Key: SULQOTGFPPFUMG-UHFFFAOYSA-N
SMILES: ClC1=C(Cl)N=C(Br)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results