accessibility menu, dialog, popup

UserName

CAS RN 16075-68-6

IUPAC Name: 6-methyl-2H-pyrido[1,2-a]pyrimidin-2-one
Synonyms: 6-methylpyrido[1,2-a]pyrimidin-2-one
Molecular Weight (g/mol): 160.18
Molecular Formula: C9H8N2O
InChi Key: XVVOALIRSDLWJA-UHFFFAOYSA-N
SMILES: CC1=CC=CC2=NC(=O)C=CN12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results