accessibility menu, dialog, popup

UserName

CAS RN 16051-77-7

IUPAC Name: (3R,3aS,6S,6aR)-6-hydroxy-hexahydrofuro[3,2-b]furan-3-yl nitrate
Synonyms: isosorbide mononitrate ismn apogepha ismn heumann monosorbitrate pentacard monis monit elantan long monicor monopront
Molecular Weight (g/mol): 191.14
Molecular Formula: C6H9NO6
InChi Key: YWXYYJSYQOXTPL-SLPGGIOYSA-N
SMILES: O[C@H]1CO[C@@H]2[C@@H](CO[C@H]12)O[N+]([O-])=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results