missing translation for 'onlineSavingsMsg'
Learn More

Isosorbide mononitrate, 98+%

Catalog No. AAJ6367414
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6367414 25 g
AAJ6367406 5 g
AAJ6367409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6367414 Supplier Thermo Scientific Chemicals Supplier No. J6367414

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A coronary vasodilator that does not require hepatic biotransformation | CAS: 16051-77-7 | C6H9NO6 | 191.139 g/mol

A coronary vasodilator that does not require hepatic biotransformation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16051-77-7
Molecular Formula C6H9NO6
Molecular Weight (g/mol) 191.139
MDL Number MFCD00143462
InChI Key YWXYYJSYQOXTPL-SLPGGIOYSA-N
Synonym isosorbide mononitrate, isosorbide 5-mononitrate, monosorbitrate, imdur, elantan, monoket, mononit, corangin, ismn, isosorbide 5-nitrate
PubChem CID 27661
ChEBI CHEBI:6062
IUPAC Name [(3S,3aR,6R,6aS)-3-hydroxy-2,3,3a,5,6,6a-hexahydrofuro[3,2-b]furan-6-yl] nitrate
SMILES C1C(C2C(O1)C(CO2)O[N+](=O)[O-])O

Specifications

Melting Point 92°C
Color White
Quantity 25 g
UN Number UN3251
Merck Index 14,5225
Formula Weight 191.14
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material Isosorbide mononitrate
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H228-H302
Flammable solid.
Harmful if swallowed.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: ISOSORBIDE-5-MONONITRATE
EINECSNumber 240-197-2
RTECSNumber LZ4386500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only