accessibility menu, dialog, popup

UserName

CAS RN 1605-18-1

IUPAC Name: 1,4-bis(prop-1-en-2-yl)benzene
Synonyms: 1,4-bis(prop-1-en-2-yl)benzene
Molecular Weight (g/mol): 158.24
Molecular Formula: C12H14
InChi Key: ZENYUPUKNXGVDY-UHFFFAOYSA-N
SMILES: CC(=C)C1=CC=C(C=C1)C(C)=C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results