CAS RN 160446-22-0
IUPAC Name:
methyl 4-(methanesulfonylmethyl)benzoate
Synonyms:
methyl 4-(methanesulfonylmethyl)benzoate
Molecular Weight (g/mol):
228.26
Molecular Formula:
C10H12O4S
InChi Key:
XCKJFFWCTKJUKD-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=C(CS(C)(=O)=O)C=C1
1
–
1
of
1
results
1
Promotions Available
Methyl 4-(methylsulfonylmethyl)benzoate, 98%
Methyl 4-(methylsulfonylmethyl)benzoate, 98%
CAS: 160446-22-0 Molecular Formula: C10H12O4S Molecular Weight (g/mol): 228.262 MDL Number: MFCD00051693 InChI Key: XCKJFFWCTKJUKD-UHFFFAOYSA-N Synonym: methyl 4-methylsulfonylmethyl benzoate,methyl 4-methanesulfonylmethyl benzoate,methyl 4-methylsulfonyl methyl benzoate,benzoic acid, 4-methylsulfonyl methyl-, methyl ester,acmc-20akgt,maybridge1_002811,methyl 4-methanesulphonylmethyl benzoate PubChem CID: 2804364 IUPAC Name: methyl 4-(methylsulfonylmethyl)benzoate SMILES: COC(=O)C1=CC=C(C=C1)CS(=O)(=O)C