missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(methylsulfonylmethyl)benzoate, 98%

Catalog No. AAL0565709
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0565709 10 g
AAL0565704 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0565709 Supplier Thermo Scientific Chemicals Supplier No. L0565709
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 160446-22-0 | C10H12O4S | 228.262 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 160446-22-0
Molecular Formula C10H12O4S
Molecular Weight (g/mol) 228.262
MDL Number MFCD00051693
InChI Key XCKJFFWCTKJUKD-UHFFFAOYSA-N
Synonym methyl 4-methylsulfonylmethyl benzoate, methyl 4-methanesulfonylmethyl benzoate, methyl 4-methylsulfonyl methyl benzoate, benzoic acid, 4-methylsulfonyl methyl-, methyl ester, acmc-20akgt, maybridge1_002811, methyl 4-methanesulphonylmethyl benzoate
PubChem CID 2804364
IUPAC Name methyl 4-(methylsulfonylmethyl)benzoate
SMILES COC(=O)C1=CC=C(C=C1)CS(=O)(=O)C

Specifications

Melting Point 164°C to 166°C
Quantity 10 g
Beilstein 7810971
Formula Weight 228.27
Percent Purity 98%
Chemical Name or Material Methyl 4-(methylsulfonylmethyl)benzoate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only