accessibility menu, dialog, popup

UserName

CH3OHOHN(R)S(R)OHN(S)(S)(S)ONHCH3H3CH3CCH3SOHOO

CAS RN 159989-65-8

IUPAC Name: (3S,4aS,8aS)-N-tert-butyl-2-[(2R,3R)-2-hydroxy-3-[(3-hydroxy-2-methylphenyl)formamido]-4-(phenylsulfanyl)butyl]-decahydroisoquinoline-3-carboxamide; methanesulfonic acid
Synonyms: methanesulfonic acid; nelfinavir
Molecular Weight (g/mol): 663.89
Molecular Formula: C33H49N3O7S2
InChi Key: NQHXCOAXSHGTIA-SKXNDZRYSA-N
SMILES: CS(O)(=O)=O.CC1=C(O)C=CC=C1C(=O)N[C@@H](CSC1=CC=CC=C1)[C@H](O)CN1C[C@H]2CCCC[C@H]2C[C@H]1C(=O)NC(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)