missing translation for 'onlineSavingsMsg'
Learn More

Nelfinavir Mesylate Hydrate 97.0+%, TCI America™

Catalog No. N098625MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N098625MG 25 mg
N0986100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N098625MG Supplier TCI America Supplier No. N098625MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 159989-65-8
Molecular Formula C33H49N3O7S2
Molecular Weight (g/mol) 663.889
MDL Number MFCD00931436
InChI Key NQHXCOAXSHGTIA-SKXNDZRYSA-N
Synonym nelfinavir mesylate, viracept, nelfinavir mesilate, nelfinavir mesylate hydrate, nelfinavir mesylate usan, unii-98d603vp8v, c32h45n3o4s.ch4o3s, nelfinavir methanesulfonate hydrate, nfv, dsstox_cid_10777
PubChem CID 64142
ChEBI CHEBI:7497
IUPAC Name (3S,4aS,8aS)-N-tert-butyl-2-[(2R,3R)-2-hydroxy-3-[(3-hydroxy-2-methylbenzoyl)amino]-4-phenylsulfanylbutyl]-3,4,4a,5,6,7,8,8a-octahydro-1H-isoquinoline-3-carboxamide;methanesulfonic acid
SMILES CC1=C(C=CC=C1O)C(=O)NC(CSC2=CC=CC=C2)C(CN3CC4CCCCC4CC3C(=O)NC(C)(C)C)O.CS(=O)(=O)O

Specifications

Color White-Yellow
Quantity 25 mg
Formula Weight 663.89
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Nelfinavir Mesylate Hydrate
RTECSNumber NW6995333
TSCA No
Recommended Storage Refrigerator