accessibility menu, dialog, popup

UserName

CAS RN 159611-02-6

OO(S)ONHOO

CAS RN 159611-02-6

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(furan-2-yl)propanoate
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(furan-2-yl)propanoate
Molecular Weight (g/mol): 376.39
Molecular Formula: C22H18NO5
InChi Key: AJXDCHXGNUFBRC-FQEVSTJZSA-M
SMILES: [O-]C(=O)[C@H](CC1=CC=CO1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results