accessibility menu, dialog, popup

UserName

CAS RN 1596-13-0

IUPAC Name: 2-cyclohexyl-5-methylphenol
Synonyms: 2-cyclohexyl-5-methylphenol
Molecular Weight (g/mol): 190.29
Molecular Formula: C13H18O
InChi Key: SRGATTGYDONWOU-UHFFFAOYSA-N
SMILES: CC1=CC(O)=C(C=C1)C1CCCCC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results