missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohexyl-5-methylphenol 97.0+%, TCI America™

Catalog No. C127210G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C127210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C127210G Supplier TCI America Supplier No. C127210G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1596-13-0
Molecular Formula C13H18O
Molecular Weight (g/mol) 190.286
MDL Number MFCD00142903
InChI Key SRGATTGYDONWOU-UHFFFAOYSA-N
Synonym 4-Cyclohexyl-3-hydroxytoluene
PubChem CID 1538498
IUPAC Name 2-cyclohexyl-5-methylphenol
SMILES CC1=CC(=C(C=C1)C2CCCCC2)O

Specifications

Melting Point 68°C
Color White-Yellow
Boiling Point 135°C
Quantity 10 g
Formula Weight 190.29
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyclohexyl-5-methylphenol
EINECSNumber (4)-1638
TSCA No