accessibility menu, dialog, popup

UserName

CAS RN 15945-07-0

IUPAC Name: 2,4,5-trichlorobenzene-1-sulfonyl chloride
Synonyms: 2,4,5-trichlorobenzenesulfonyl chloride
Molecular Weight (g/mol): 279.94
Molecular Formula: C6H2Cl4O2S
InChi Key: WNVVRCKTQSCPAC-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(C=C1Cl)S(Cl)(=O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)