missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trichlorobenzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. B0247500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0247500G 500 g
B024725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0247500G Supplier TCI America Supplier No. B0247500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15945-07-0
Molecular Formula C6H2Cl4O2S
Molecular Weight (g/mol) 279.94
MDL Number MFCD00007428
InChI Key WNVVRCKTQSCPAC-UHFFFAOYSA-N
Synonym 2,4,5-trichlorobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 2,4,5-trichloro, unii-7gd5283xix, 2,4,5-trichlorobenzenesulphonyl chloride, 2,4,5-trichlorobenzenesulfonylchloride, pubchem5174, acmc-1c50t, 2,4,5trichlorophenylsulfonylchloride, 2,5-trichlorobenzenesulfonyl chloride, chloro 2,4,5-trichlorophenyl sulfone
PubChem CID 27575
IUPAC Name 2,4,5-trichlorobenzenesulfonyl chloride
SMILES C1=C(C(=CC(=C1Cl)Cl)Cl)S(=O)(=O)Cl

Specifications

Melting Point 68°C
Color White-Yellow
Quantity 500 g
UN Number 3261
Formula Weight 279.94
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,5-Trichlorobenzenesulfonyl Chloride