CAS RN 159351-69-6
IUPAC Name:
(1R,9S,12S,15R,16Z,18R,19R,21R,23S,24E,26Z,28E,30S,32S,35R)-1,18-dihydroxy-12-[(2R)-1-[(1S,3R,4R)-4-(2-hydroxyethoxy)-3-methoxycyclohexyl]propan-2-yl]-19,30-dimethoxy-15,17,21,23,29,35-hexamethyl-11,36-dioxa-4-azatricyclo[30.3.1.0⁴,⁹]hexatriaconta-16,24,26,28-tetraene-2,3,10,14,20-pentone
Synonyms:
everolimus sdz rad RAD everolimus (substance) certican sdz-rad everolimus (product)
Molecular Weight (g/mol):
958.24
Molecular Formula:
C53H83NO14
InChi Key:
HKVAMNSJSFKALM-CNPAPGRKSA-N
SMILES:
CO[C@@H]1C[C@H](C[C@@H](C)[C@@H]2CC(=O)[C@H](C)\C=C(C)/[C@@H](O)[C@@H](OC)C(=O)[C@H](C)C[C@H](C)\C=C\C=C/C=C(C)/[C@H](C[C@@H]3CC[C@@H](C)[C@@](O)(O3)C(=O)C(=O)N3CCCC[C@H]3C(=O)O2)OC)CC[C@H]1OCCO
1
–
3
of
3
results
1
Everolimus
CAS: 159351-69-6 Molecular Formula: C53H83NO14 Molecular Weight (g/mol): 958.24 MDL Number: MFCD07785165 InChI Key: HKVAMNSJSFKALM-PWMUVMERSA-N PubChem CID: 129317877 SMILES: CC1CCC2CC(C(=CC=CC=CC(CC(C(=O)C(C(C(=CC(C(=O)CC(OC(=O)C3CCCCN3C(=O)C(=O)C1(O2)O)C(C)CC4CCC(C(C4)OC)OCCO)C)C)O)OC)C)C)C)OC
2
Everolimus Analytical Standard, MilliporeSigma™ Supelco™
A rapamycin derivative that inhibits the mTOR pathway, useful as an analytical standard for clinical research.
3
R&D Systems™ Everolimus
A Tocris product, part of R&D Systems. mTOR inhibitor;immunosuppressant