missing translation for 'onlineSavingsMsg'
Learn More

Everolimus

Catalog No. AAJ60139MB
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60139MB 25 mg
AAJ60139LB0 5 mg
AAJ60139MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ60139MB Supplier Thermo Scientific Chemicals Supplier No. J60139MB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A rapamycin analog | CAS: 159351-69-6 | C53H83NO14 | 958.24 g/mol

Everolimus is a rapamycin analog. It is used to treat certain types of kidney cancer, breast cancer and brain tumor. It prevents transplant rejection by decreasing the activity of the immune system. It also act as a kinase inhibitor. It is used in the treatment of subependymal giant cell astrocytoma (SEGA; a type of brain tumor) in adults and children 1 year of age and older who have TSC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in dimethysulfoxide,ethanol and chloroform. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 159351-69-6
Molecular Formula C53H83NO14
Molecular Weight (g/mol) 958.24
MDL Number MFCD07785165
InChI Key HKVAMNSJSFKALM-PWMUVMERSA-N
PubChem CID 129317877
SMILES CC1CCC2CC(C(=CC=CC=CC(CC(C(=O)C(C(C(=CC(C(=O)CC(OC(=O)C3CCCCN3C(=O)C(=O)C1(O2)O)C(C)CC4CCC(C(C4)OC)OCCO)C)C)O)OC)C)C)C)OC

Specifications

Color White
Quantity 25 mg
Merck Index 14,3907
Solubility Information Soluble in dimethysulfoxide,ethanol and chloroform; Slightly soluble in water.
Formula Weight 958.22
Physical Form Powder
Chemical Name or Material Everolimus
Hazard Category H372
Hazard Statement GHS H Statement
H372
Causes damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P314-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only