missing translation for 'onlineSavingsMsg'
Learn More

Everolimus Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022814
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022814 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022814 Supplier Merck Emd Millipore Supplier No. 9468710MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A rapamycin derivative that inhibits the mTOR pathway, useful as an analytical standard for clinical research.

Specifications

CAS 159351-69-6
Flash Point Not Applicable
Molecular Formula C53H83NO14
Linear Formula C53H83NO14
Quantity 10 mg
MDL Number MFCD00929329
Synonym 42-O-(2-Hydroxyethyl)rapamycin
Molecular Weight (g/mol) 958.22
Formula Weight 958.22
Percent Purity ≥75% (qNMR)
Grade Analytical Standard
Physical Form Solid
Show More Show Less