CAS RN 15898-43-8
IUPAC Name:
4-acetamidobenzene-1-sulfinic acid sodium
Synonyms:
4-acetamidobenzenesulfinic acid sodium
Molecular Weight (g/mol):
222.21
Molecular Formula:
C8H9NNaO3S
InChi Key:
GFNIDYGMWDLXEY-UHFFFAOYNA-N
SMILES:
[Na].CC(=O)NC1=CC=C(C=C1)S(O)=O
1
–
1
of
1
results
1
Sodium 4-Acetamidobenzenesulfinate Dihydrate 98.0+%, TCI America™
CAS: 15898-43-8 Molecular Formula: C8H12NNaO5S Molecular Weight (g/mol): 257.236 MDL Number: MFCD04113633 InChI Key: LKRSHPGOHITHSF-UHFFFAOYSA-M Synonym: 4-Acetamidobenzenesulfinic Acid Sodium Salt PubChem CID: 70699978 IUPAC Name: sodium;4-acetamidobenzenesulfinate;dihydrate SMILES: CC(=O)NC1=CC=C(C=C1)S(=O)[O-].O.O.[Na+]