missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-Acetamidobenzenesulfinate Dihydrate 98.0+%, TCI America™

Catalog No. A00115G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A00115G 5 g
A001125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A00115G Supplier TCI America Supplier No. A00115G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15898-43-8
Molecular Formula C8H12NNaO5S
Molecular Weight (g/mol) 257.236
MDL Number MFCD04113633
InChI Key LKRSHPGOHITHSF-UHFFFAOYSA-M
Synonym 4-Acetamidobenzenesulfinic Acid Sodium Salt
PubChem CID 70699978
IUPAC Name sodium;4-acetamidobenzenesulfinate;dihydrate
SMILES CC(=O)NC1=CC=C(C=C1)S(=O)[O-].O.O.[Na+]

Specifications

Color White
Quantity 5 g
Formula Weight 221.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 4-Acetamidobenzenesulfinate Dihydrate
EINECSNumber (3)-1987
TSCA No