accessibility menu, dialog, popup

UserName

CAS RN 158932-00-4

IUPAC Name: tert-butyl N-[(2R)-1-hydroxy-3-(1H-indol-3-yl)propan-2-yl]carbamate
Synonyms: tert-butyl N-[(2R)-1-hydroxy-3-(1H-indol-3-yl)propan-2-yl]carbamate|BOC-D-TRYPTOPHANOL
Molecular Weight (g/mol): 290.36
Molecular Formula: C16H22N2O3
InChi Key: JEFQUFUAEKORKL-GFCCVEGCSA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CO)CC1=CNC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results