missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N(alpha)-Boc-D-tryptophanol, 98%

Catalog No. AAH2776303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2776303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2776303 Supplier Thermo Scientific Chemicals Supplier No. H2776303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 158932-00-4
Molecular Formula C16H22N2O3
Molecular Weight (g/mol) 290.36
MDL Number MFCD00270221
InChI Key JEFQUFUAEKORKL-GFCCVEGCSA-N
Synonym boc-d-tryptophanol, n-boc-d-tryptophanol, r-tert-butyl 1-hydroxy-3-1h-indol-3-yl propan-2-yl carbamate, n alpha-boc-d-tryptophanol, boc-d-trp-ol, tert-butyl n-2r-1-hydroxy-3-1h-indol-3-yl propan-2-yl carbamate, ambotzbal1034, n-t-boc-d-tryptophanol, tert-butyl r-1-hydroxy-3-1h-indol-3-yl propan-2-yl carbamate, tert-butyl 2r-1-hydroxy-3-1h-indol-3-yl propan-2-yl carbamate
PubChem CID 20744818
SMILES CC(C)(C)OC(=O)N[C@@H](CO)CC1=CNC2=CC=CC=C12

Specifications

Melting Point 119°C to 121°C
Quantity 1 g
Optical Rotation +53° (c=2 in Acetic Acid)
IUPAC Name tert-butyl N-[(2R)-1-hydroxy-3-(1H-indol-3-yl)propan-2-yl]carbamate
Formula Weight 290.36
Percent Purity 98%
Chemical Name or Material N(alpha)-Boc-D-tryptophanol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only