accessibility menu, dialog, popup

UserName

CAS RN 158871-28-4

IUPAC Name: 3-({5-[(2-cyanoethyl)sulfanyl]-2-oxo-2H-1,3-dithiol-4-yl}sulfanyl)propanenitrile
Synonyms: 3-({5-[(2-cyanoethyl)sulfanyl]-2-oxo-1,3-dithiol-4-yl}sulfanyl)propanenitrile
Molecular Weight (g/mol): 288.42
Molecular Formula: C9H8N2OS4
InChi Key: SIYHFUIJFJOCQE-UHFFFAOYSA-N
SMILES: O=C1SC(SCCC#N)=C(SCCC#N)S1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results