missing translation for 'onlineSavingsMsg'
Learn More

4,5-Bis(2-cyanoethylthio)-1,3-dithiol-2-one 98.0+%, TCI America™

Catalog No. B22331G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B22331G Supplier TCI America Supplier No. B22331G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 158871-28-4
Molecular Formula C9H8N2OS4
Molecular Weight (g/mol) 288.42
MDL Number MFCD01142888
InChI Key SIYHFUIJFJOCQE-UHFFFAOYSA-N
PubChem CID 4260353
IUPAC Name 3-({5-[(2-cyanoethyl)sulfanyl]-2-oxo-2H-1,3-dithiol-4-yl}sulfanyl)propanenitrile
SMILES O=C1SC(SCCC#N)=C(SCCC#N)S1

Specifications

Melting Point 84°C
Quantity 1 g
UN Number 3439
Formula Weight 288.42
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Bis(2-cyanoethylthio)-1,3-dithiol-2-one