accessibility menu, dialog, popup

UserName

HOHO(R)OO(R)O(R)(S)HOClClCl

CAS RN 15879-93-3

IUPAC Name: 1-[(2R,5R,6S,6aR)-6-hydroxy-2-(trichloromethyl)-tetrahydro-2H-furo[2,3-d][1,3]dioxol-5-yl]ethane-1,2-diol
Synonyms: chloralose
Molecular Weight (g/mol): 309.52
Molecular Formula: C8H11Cl3O6
InChi Key: OJYGBLRPYBAHRT-OPKHMCHVSA-N
SMILES: OCC(O)[C@H]1OC2O[C@@H](O[C@@H]2[C@H]1O)C(Cl)(Cl)Cl
Quantity 
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)