accessibility menu, dialog, popup

UserName

OHNNOOBr

CAS RN 15862-33-6

IUPAC Name: 3-bromo-5-nitro-1,2-dihydropyridin-2-one
Synonyms: 3-bromo-5-nitro-1H-pyridin-2-one
Molecular Weight (g/mol): 218.99
Molecular Formula: C5H3BrN2O3
InChi Key: KWSOHRDMTWDAOI-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CNC(=O)C(Br)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results