accessibility menu, dialog, popup

UserName

NHN

CAS RN 15861-24-2

IUPAC Name: 1H-indole-5-carbonitrile
Synonyms: indole-5-cyano-
Molecular Weight (g/mol): 142.16
Molecular Formula: C9H6N2
InChi Key: YHYLDEVWYOFIJK-UHFFFAOYSA-N
SMILES: N#CC1=CC=C2NC=CC2=C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (5)