accessibility menu, dialog, popup

UserName

CAS RN 1583-58-0

IUPAC Name: 2,4-difluorobenzoic acid
Synonyms: 2,4-difluorobenzoic acid
Molecular Weight (g/mol): 158.1
Molecular Formula: C7H4F2O2
InChi Key: NJYBIFYEWYWYAN-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(F)C=C1F
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)