missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzoic Acid 98.0+%, TCI America™

Catalog No. D1833500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1833500G 500 g
D183325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1833500G Supplier TCI America Supplier No. D1833500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1583-58-0
Molecular Formula C7H4F2O2
Molecular Weight (g/mol) 158.10
MDL Number MFCD00011670
InChI Key NJYBIFYEWYWYAN-UHFFFAOYSA-N
Synonym 2,4-dfba, benzoic acid, 2,4-difluoro, 2,4-difluorobenzoicacid, 2,4-difluoro-benzoic acid, zlchem 472, pubchem1336, acmc-1brgf, 2,4-difluorbenzoic acid, 2,4-difluorobezoic acid, 2 4-difluorobenzoic acid
PubChem CID 74102
IUPAC Name 2,4-difluorobenzoic acid
SMILES OC(=O)C1=CC=C(F)C=C1F

Specifications

Melting Point 188°C
Quantity 500 g
Formula Weight 158.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Difluorobenzoic Acid