accessibility menu, dialog, popup

UserName

CAS RN 1572-84-5

CH3OOOOCH3(E/Z)

CAS RN 1572-84-5

IUPAC Name: 4-(butanoyloxy)but-2-en-1-yl butanoate
Synonyms: 4-(butanoyloxy)but-2-en-1-yl butanoate
Molecular Weight (g/mol): 228.29
Molecular Formula: C12H20O4
InChi Key: OGJKSGMVEHGEKF-UHFFFAOYSA-N
SMILES: CCCC(=O)OCC=CCOC(=O)CCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results