missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Bis(butyryloxy)-2-butene 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 155°C |
| Quantity | 5 g |
| Formula Weight | 228.29 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1,4-Bis(butyryloxy)-2-butene |
Safety and Handling
| TSCA | No |