accessibility menu, dialog, popup

UserName

OHONNN

CAS RN 157069-48-2

IUPAC Name: 4-(4H-1,2,4-triazol-4-yl)benzoic acid
Synonyms: 4-(1,2,4-triazol-4-yl)benzoic acid
Molecular Weight (g/mol): 189.17
Molecular Formula: C9H7N3O2
InChi Key: QCNDLFQKKMPZKH-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)N1C=NN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)