missing translation for 'onlineSavingsMsg'
Learn More

4-(p-Carboxyphenyl)-1,2,4-triazole, 97%, Thermo Scientific™

Catalog No. p-7077332
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC449960010 1g
AC449960050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC449960010 Supplier Thermo Scientific Chemicals Supplier No. 449960010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 157069-48-2
Molecular Formula C9H7N3O2
Molecular Weight (g/mol) 189.17
InChI Key QCNDLFQKKMPZKH-UHFFFAOYSA-N
Synonym 4-4h-1,2,4-triazol-4-yl benzoic acid, 4-1,2,4-triazol-4-yl benzoic acid, 4-1,2,4 triazol-4-yl-benzoic acid, benzoic acid, 4-4h-1,2,4-triazol-4-yl, 4-4-carboxyphenyl-1,2,4-triazole, 4-4-carboxyphenyl-4h-1,2,4-triazole, 4-4h-1,2,4-triazol-4-yl-benzoic acid
PubChem CID 3163581
IUPAC Name 4-(1,2,4-triazol-4-yl)benzoic acid
SMILES C1=CC(=CC=C1C(=O)O)N2C=NN=C2

Specifications

Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1g
Formula Weight 189.17
Percent Purity 97%
Chemical Name or Material 4-(p-Carboxyphenyl)-1,2,4-triazole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only