CAS RN 1569-69-3
CAS RN 1569-69-3
IUPAC Name:
cyclohexanethiol
Synonyms:
cyclohexanethiol
Molecular Weight (g/mol):
116.22
Molecular Formula:
C6H12S
InChi Key:
CMKBCTPCXZNQKX-UHFFFAOYSA-N
SMILES:
SC1CCCCC1
1
–
1
of
1
results
1
Cyclohexanethiol 98.0+%, TCI America™
CAS: 1569-69-3 Molecular Formula: C6H12S Molecular Weight (g/mol): 116.222 MDL Number: MFCD00001455 InChI Key: CMKBCTPCXZNQKX-UHFFFAOYSA-N Synonym: cyclohexyl mercaptan,cyclohexyl thiol,cyclohexylmercaptan,cyclohexylthiol,cyklohexylmerkaptan,cyklohexanthiol,cyklohexanthiol czech,cyklohexylmerkaptan czech,hexahydrothiophenol,mercaptocyclohexane PubChem CID: 15290 IUPAC Name: cyclohexanethiol SMILES: C1CCC(CC1)S