missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexanethiol 98.0+%, TCI America™
Spécifications
Spécifications
| Color | Colorless |
| Boiling Point | 158°C |
| Quantity | 500 mL |
| UN Number | 3054 |
| Formula Weight | 116.22 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | Cyclohexanethiol |
Sécurité et manipulation
| EINECSNumber | (3)-2303 |
| RTECSNumber | GV7525000 |
| TSCA | Yes |