accessibility menu, dialog, popup

UserName

CH3H3CCH3OOHN(S)OHO

CAS RN 155976-13-9

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-2-cyclopropylacetic acid
Synonyms: (S)-[(tert-butoxycarbonyl)amino](cyclopropyl)acetic acid
Molecular Weight (g/mol): 215.25
Molecular Formula: C10H17NO4
InChi Key: QFVJNEASAAJIDF-ZETCQYMHSA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](C1CC1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)