missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-2-cyclopropyl-L-glycine, 95%, Thermo Scientific Chemicals

Catalog No. AAH6291403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6291403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6291403 Supplier Thermo Scientific Chemicals Supplier No. H6291403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 155976-13-9
Molecular Formula C10H17NO4
Molecular Weight (g/mol) 215.25
MDL Number MFCD06659132,MFCD01861823
InChI Key QFVJNEASAAJIDF-ZETCQYMHSA-N
Synonym boc-l-cyclopropylglycine, s-2-tert-butoxycarbonyl amino-2-cyclopropylacetic acid, boc-s-cyclopropylglycine, s-tert-butoxycarbonylamino-cyclopropyl-acetic acid, s-boc-amino-cyclopropylacetic acid, 2s-2-amino-2-cyclopropylethanoic acid, n-boc protected, s-2-tert-butoxycarbonylamino-2-cyclopropylacetic acid, s-tert-butoxycarbonyl amino cyclopropyl acetic acid, boc-l-cyclopropyl glycine
PubChem CID 2761461
IUPAC Name (2S)-2-cyclopropyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid
SMILES CC(C)(C)OC(=O)N[C@@H](C1CC1)C(O)=O

Specifications

Color White
Beilstein 6650020
Quantity 1 g
Formula Weight 215.25
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Boc-2-cyclopropyl-L-glycine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only