CAS RN 1558-17-4
IUPAC Name:
4,6-dimethylpyrimidine
Synonyms:
4,6-dimethylpyrimidine
Molecular Weight (g/mol):
108.14
Molecular Formula:
C6H8N2
InChi Key:
LSBIUXKNVUBKRI-UHFFFAOYSA-N
SMILES:
CC1=CC(C)=NC=N1
1
–
1
of
1
results
1
4,6-Dimethylpyrimidine 98.0+%, TCI America™
CAS: 1558-17-4 Molecular Formula: C6H8N2 Molecular Weight (g/mol): 108.14 MDL Number: MFCD00006116 InChI Key: LSBIUXKNVUBKRI-UHFFFAOYSA-N Synonym: 4,6-dimethyl-pyrimidine,pyrimidine, 4,6-dimethyl,unii-95n99d3vhb,pyrimidine, 4,6-dimethyl-6ci,7ci,8ci,9ci,dimethylpyrimidinetech,zlchem 778,pubchem21498,4,6-dimethyl-pyrimidin,4,6-dimethyl-pyrmidine,acmc-1bwo9 PubChem CID: 15257 IUPAC Name: 4,6-dimethylpyrimidine SMILES: CC1=CC(C)=NC=N1