missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dimethylpyrimidine 98.0+%, TCI America™

Catalog No. D42825G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42825G 5 g
D42821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42825G Supplier TCI America Supplier No. D42825G

Chemical Identifiers

CAS 1558-17-4
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00006116
InChI Key LSBIUXKNVUBKRI-UHFFFAOYSA-N
Synonym 4,6-dimethyl-pyrimidine, pyrimidine, 4,6-dimethyl, unii-95n99d3vhb, pyrimidine, 4,6-dimethyl-6ci,7ci,8ci,9ci, dimethylpyrimidinetech, zlchem 778, pubchem21498, 4,6-dimethyl-pyrimidin, 4,6-dimethyl-pyrmidine, acmc-1bwo9
PubChem CID 15257
IUPAC Name 4,6-dimethylpyrimidine
SMILES CC1=CC(C)=NC=N1

Specifications

Color White
Boiling Point 159°C
Quantity 5 g
UN Number 1325
Formula Weight 108.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4,6-Dimethylpyrimidine