accessibility menu, dialog, popup

UserName

CAS RN 1552-96-1

IUPAC Name: (2E)-3-[4-(dimethylamino)phenyl]prop-2-enoic acid
Synonyms: (2E)-3-[4-(dimethylamino)phenyl]prop-2-enoic acid
Molecular Weight (g/mol): 191.23
Molecular Formula: C11H13NO2
InChi Key: CQNPVMCASGWEHM-VMPITWQZSA-N
SMILES: CN(C)C1=CC=C(\C=C\C(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results