missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminocinnamic acid, 97+%

Catalog No. AAB2154906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2154906 5 g
AAB2154903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2154906 Supplier Thermo Scientific Chemicals Supplier No. B2154906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1552-96-1 | C11H13NO2 | 191.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1552-96-1
Molecular Formula C11H13NO2
Molecular Weight (g/mol) 191.23
MDL Number MFCD00004397
InChI Key CQNPVMCASGWEHM-VMPITWQZSA-N
Synonym 4-dimethylamino cinnamic acid, 4-dimethylaminocinnamic acid, p-dimethylamino cinnamic acid, 3-4-dimethylamino phenyl acrylic acid, 4-n,n-dimethylamino cinnamic acid, 2e-3-4-dimethylamino phenyl prop-2-enoic acid, e-3-4-dimethylaminophenyl prop-2-enoic acid, e-3-4-dimethylamino phenyl acrylic acid
PubChem CID 1540638
IUPAC Name (E)-3-[4-(dimethylamino)phenyl]prop-2-enoic acid
SMILES CN(C)C1=CC=C(C=C1)C=CC(=O)O

Specifications

Melting Point ∼222°C (decomposition)
Quantity 5 g
Beilstein 1368533
Formula Weight 191.23
Percent Purity 99%
Chemical Name or Material 4-Dimethylaminocinnamic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 216-299-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only